missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFD, Mouse anti-Human, Clone: 4H2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00080310-M09.100ug
This item is not returnable.
View return policy
Description
Sequence: TPQSASIKALRNANLRRDDLYRRDETIQVKGNGYVQSPRFPNSYPRNLLLTWRLHSQENTRIQLVFDNQFGLEEAENDICRYDFVEVEDISETSTIIRGRSpecifications
| PDGFD | |
| Monoclonal | |
| Unconjugated | |
| platelet derived growth factor D | |
| IEGF/MGC26867/MSTP036/SCDGF-B/SCDGFB | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunoprecipitation | |
| 4H2 | |
| In 1x PBS, pH 7.4 | |
| NM_033135 | |
| PDGFD | |
| PDGFD (NP_149126, 24 a.a. ∼ 123 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 80310 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction