missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFR alpha Antibody (16A1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-44581CL1
This item is not returnable.
View return policy
Description
PDGFR alpha Monoclonal antibody specifically detects PDGFR alpha in Human samples. It is validated for Flow Cytometry,In vitro Assay,Flow (Cell Surface),CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| PDGFR alpha | |
| Monoclonal | |
| CoraFluor 1 | |
| alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
| CD140a-transfected NIH 3T3 cells | |
| 0.1 mL | |
| Phospho Specific, Protein Kinase | |
| 5156 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, In vitro Assay, Flow Cytometry (Cell Surface), CyTOF, Immunocytochemistry/Immunofluorescence | |
| 16A1 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction