missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFR alpha Antibody (Hu136) [Allophycocyanin], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals FAB10352A
This item is not returnable.
View return policy
Description
PDGFR alpha Monoclonal antibody specifically detects PDGFR alpha in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| PDGFR alpha | |
| Monoclonal | |
| APC | |
| PBS | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| Hu136 | |
| Flow Cytometry, CyTOF-ready | |
| alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
| Human PDGF R alpha | |
| 0.1 mL | |
| Protein Kinase | |
| 5156 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction