missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFR alpha Antibody (MM0004-8A89), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60969DL594
This item is not returnable.
View return policy
Description
PDGFR alpha Monoclonal antibody specifically detects PDGFR alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PDGFR alpha | |
| Monoclonal | |
| DyLight 594 | |
| alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
| Human recombinant PDGFR alpha (N-terminal) | |
| 0.1 mL | |
| Protein Kinase | |
| 5156 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0004-8A89 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction