missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFR alpha Antibody (PRa292) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1264JF525
This item is not returnable.
View return policy
Description
PDGFR alpha Monoclonal antibody specifically detects PDGFR alpha in Human samples. It is validated for Flow Cytometry,Immunoprecipitation
Specifications
| PDGFR alpha | |
| Monoclonal | |
| Janelia Fluor 525 | |
| alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
| Human osteosarcoma cell membrane extracts | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunoprecipitation | |
| PRa292 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.25 mL | |
| Phospho Specific, Protein Kinase | |
| 5156 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction