missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFRB, Mouse anti-Human, Clone: 4C12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005159-M08.100ug
This item is not returnable.
View return policy
Description
Sequence: LVVTPPGPELVLNVSSTFVLTCSGSAPVVWERMSQEPPQEMAKAQDGTFSSVLTLTNLTGLDTGEYFCTHNDSRGLETDERKRLYIFVPDPTVGFLPNDAESpecifications
| PDGFRB | |
| Monoclonal | |
| Unconjugated | |
| platelet-derived growth factor receptor, beta polypeptide | |
| CD140B/JTK12/PDGF-R-beta/PDGFR/PDGFR1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunoprecipitation, In situ PLA, Western Blot | |
| 4C12 | |
| In 1x PBS, pH 7.4 | |
| BC032224 | |
| PDGFRB | |
| PDGFRB (AAH32224, 33 a.a. ∼ 133 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5159 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction