missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFRL Antibody (1008804) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1723I
This item is not returnable.
View return policy
Description
PDGFRL Monoclonal antibody specifically detects PDGFRL in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| PDGFRL | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 1008804 | |
| Flow Cytometry, CyTOF-ready | |
| PDGFR-like protein, PDGRL, platelet-derived growth factor receptor-like, platelet-derived growth factor receptor-like protein, platelet-derived growth factor-beta-like tumor suppressor, PRLTSPDGF receptor beta-like tumor suppressor | |
| Human embryonic kidney cell, HEK293-derived human PDGFRL, Gln22-Ser375, Accession # Q15198 | |
| 0.1 mL | |
| Cancer, GPCR, Signal Transduction | |
| 5157 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction