Learn More
PDH E1 alpha Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18068-1-AP-150UL
 View more versions of this product
Product Code. 16868104
  • € 410.00 / 150µL
Description
Pyruvate dehydrogenase (PDH) is a mitochondrial multienzyme complex that catalyzes the oxidative decarboxylation of pyruvate and is one of the major enzymes responsible for the regulation of homeostasis of carbohydrate fuels in mammals. The enzymatic activity is regulated by a phosphorylation/dephosphorylation cycle. Phosphorylation of PDH by a specific pyruvate dehydrogenase kinase (PDK) results in inactivation.
Specifications
Specifications
PDH E1 alpha
Polyclonal
Unconjugated
PDHA1
PDH E1 alpha, PDH E1alpha, PDHA1, PDHE1 A type I, pyruvate dehydrogenase E1 alpha subunit
Rabbit
Antigen Affinity Chromatography
RUO
18597, 29554, 5160
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08559, P26284, P35486
Pdha1
PDH E1alpha Fusion Protein Ag12701
150 μL
Primary
Human, Mouse, Rat