missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDHA2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94780-0.02ml
This item is not returnable.
View return policy
Description
PDHA2 Polyclonal antibody specifically detects PDHA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| PDHA2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 | |
| EC 1.2.4.1, MGC149517, MGC149518, mitochondrial, PDHAL, PDHE1-A type II, pyruvate dehydrogenase (lipoamide) alpha 2, pyruvate dehydrogenase E1 component subunit alpha, testis-specific form, pyruvate dehydrogenase, E1-alpha polypeptide, testis specific | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 119-388 of human PDHA2 (NP_005381.1). HGVCYTRGLSVRSILAELTGRRGGCAKGKGGSMHMYTKNFYGGNGIVGAQGPLGAGIALACKYKGNDEICLTLYGDGAANQGQIAEAFNMAALWKLPCVFICENNLYGMGTSTERAAASPDYYKRGNFIPGLKVDGMDVLCVREATKFAANYCRSGKGPILMELQTYRYHGHSMSDPGVSYRTREEIQEVRSKRDPIIILQDRMVNSKLATVEELKEIGAEVRKEIDDAAQFATTDPEPHLEELGHHIYSSDSSFEVRGANPWIKFKSVS | |
| 0.02 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 5161 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction