missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDHA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561531
This item is not returnable.
View return policy
Description
Immunogen sequence: ILAELTGRRG GCAKGKGGSM HMYTKNFYGG NGIVG Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
PDHA2 (Pyruvate Dehydrogenase E1 Alpha 2 Subunit) is a Protein Coding gene. Diseases associated with PDHA2 include Spermatogenic Failure 1. Among its related pathways are Citrate cycle (TCA cycle)and Central carbon metabolism in cancer. Gene Ontology (GO) annotations related to this gene include oxidoreductase activity, acting on the aldehyde or oxo group of donors, disulfide as acceptorandpyruvate dehydrogenase (acetyl-transferring) activity. An important paralog of this gene isPDHA1.
Specifications
| PDHA2 | |
| Polyclonal | |
| Unconjugated | |
| PDHA2 | |
| Pdha2; Pdha-2; Pdhal; PDHE1-A type II; pyruvate dehydrogenase; pyruvate dehydrogenase (lipoamide) alpha 2; pyruvate dehydrogenase E1 alpha 2; PYRUVATE DEHYDROGENASE E1 COMPONENT ALPHA SUBUNIT, TESTIS-SPECIFIC FORM, MITOCHONDRIAL PRECURSOR (PDHE1-A TYPE II); pyruvate dehydrogenase E1 component subunit alpha, testis-specific form, mitochondrial; pyruvate dehydrogenase, E1-alpha polypeptide, testis specific | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5161 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P29803 | |
| PDHA2 | |
| Recombinant protein corresponding to Human PDHA2. Recombinant protein control fragment (Product #RP-99528). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction