missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDHB Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA557792

Product Code. 15785681

  • € 534.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MAGLRPICEF MTFNFSMQAI DQVINSAAKT YYMSGGLQPV PIVFRGPNGA SAGVAAQHSQ CFAAWYGHCP GLKVVSPWNS EDAKG Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

PDHB (Pyruvate dehydrogenase E1 component subunit beta, mitochondrial) is also named as PHE1B. The enzyme, which is found in mitochondria, is one of the component enzymes of the pyruvate dehydrogenase multienzyme complex (PDH). It catalyzes the first reaction of an oxidative decarboxylation sequence converting pyruvate to acetyl-CoA and CO(2). Defects in PDHB are the cause of pyruvate dehydrogenase E1-beta deficiency (PDHBD).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PDHB
Polyclonal
Unconjugated
PDHB
2610103L06Rik; AL024199; C81408; PDHB; PDHBD; PDHE1-B; PHE1B; pyruvate dehydrogenase (lipoamide) beta; pyruvate dehydrogenase E1 component subunit beta, mitochondrial; pyruvate dehydrogenase, E1 beta polypeptide
Rabbit
Antigen affinity chromatography
RUO
5162
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P11177
PDHB
Recombinant protein corresponding to Human PDHB. Recombinant protein control fragment (Product #RP-96432).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.