missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDIA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35272-100ul
This item is not returnable.
View return policy
Description
PDIA2 Polyclonal antibody specifically detects PDIA2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| PDIA2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Pancreas-specific protein disulfide isomerase, pancreatic protein disulfide isomerase, PDA2, PDI, PDIp, PDIR, protein disulfide isomerase, pancreatic, protein disulfide isomerase-associated 2, protein disulfide-isomerase A2, Rho GDP dissociation inhibitor gamma | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 326-525 of human PDIA2 (NP_006840.2).,, Sequence:, FGLKAEAAPTLRLVNLETTKKYAPVDGGPVTAASITAFCHAVLNGQVKPYLLSQEIPPDWDQRPVKTLVGKNFEQVAFDETKNVFVKFYAPWCTHCKEMAPAWEALAEKYQDHEDIIIAELDATANELDAFAVHGFPTLKYFPAGPGRKVIEYKSTRDLETFSKFLDNGGVLPTEEPPEEPAAPFPEPPANSTMGSKEEL | |
| 100 μL | |
| Biologically Active Proteins, Cancer, Cellular Markers, ER Markers, Hypoxia | |
| 64714 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction