missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDIA3 Monoclonal Antibody (OTI2D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504993
This item is not returnable.
View return policy
Description
This gene encodes a protein of the endoplasmic reticulum that interacts with lectin chaperones calreticulin and calnexin to modulate folding of newly synthesized glycoproteins. The protein was once thought to be a phospholipase; however, it has been demonstrated that the protein actually has protein disulfide isomerase activity. It is thought that complexes of lectins and this protein mediate protein folding by promoting formation of disulfide bonds in their glycoprotein substrates.Specifications
| PDIA3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P30101 | |
| PDIA3 | |
| Human recombit protein fragment corresponding to amino acids 140-505 of human PDIA3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI2D2 | |
| Unconjugated | |
| PDIA3 | |
| ER60, ERp57, ERp60, ERp61, GRP57, GRP58, HEL-S-269, HEL-S-93n, HsT17083, P58, PI-PLC | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2923 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction