Learn More
PDIA3 Mouse anti-Human, CoraLite 488, Clone: 3G4G7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66423-100UL
Description
The PDIA3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PDIA3. This antibody recognizes human, mouse, rat antigen. The PDIA3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| PDIA3 | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Immunofluorescence | |
| 3G4G7 | |
| CoraLite 488 | |
| 100 μL | |
| 2923 | |
| IgG1 |