Learn More
PDIA3 Mouse anti-Human, CoraLite 488, Clone: 3G4G7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66423-100UL
Product Code. 16860796
  • € 450.00 / 100µL
Description
The PDIA3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PDIA3. This antibody recognizes human, mouse, rat antigen. The PDIA3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
PDIA3
Monoclonal
2000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Immunofluorescence
3G4G7
CoraLite 488
100 μL
2923