missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDIA4 Monoclonal Antibody (OTI6B6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503888S
This item is not returnable.
View return policy
Description
PDIA4 Monoclonal antibody specifically detects PDIA4 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| PDIA4 | |
| Monoclonal | |
| 0.87 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P13667 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9601 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI6B6 | |
| Unconjugated | |
| PDIA4 | |
| AI987846; caBP2; Cai; calcium binding protein, intestinal; calcium-binding protein 2; EC 5.3.4.1; endoplasmic reticulum protein 72; endoplasmic reticulum resident protein 70; Endoplasmic reticulum resident protein 72; ER protein 70; ER protein 72; Erp70; Erp72; ERp-72; PDIA4; protein disu; protein disulfide isomerase A4; protein disulfide isomerase associated 4; protein disulfide isomerase family A member 4; protein disulfide isomerase family A, member 4; protein disulfide isomerase related protein (calcium-binding protein, intestinal-related); protein disulfide isomerase-associated 4; protein disulfide-isomerase A4; Protein ERp-72; testicular tissue protein Li 137 | |
| Full length human recombinant protein of human PDIA4 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction