missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDIK1L Monoclonal Antibody (4B7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523088
This item is not returnable.
View return policy
Description
Peptide Sequence: MVSSQPKYDL IREVGRGSYG VVYEAVIRKT SARVAVKKIR CHAPENVELA LREFWALSSI KSQHPNVIHL EECILQKDGM VQKMSHGSNS SLYLQLVET.
PDIK1L is a protein coding gene.
Specifications
| PDIK1L | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q8N165 | |
| PDIK1L | |
| PDIK1L (NP_690048, 1 a.a. ~ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA | |
| 4B7 | |
| Unconjugated | |
| PDIK1L | |
| BC027088; casein kinase; CLIK1L; PDIK1L; PDLIM1 interacting kinase 1 like; PDLIM1-interacting kinase 1-like; Serine/threonine-protein kinase PDIK1L; STK35L2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 149420 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction