Learn More
PDIK1L Monoclonal Antibody (OTI4G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811104S
This item is not returnable.
View return policy
Description
PDIK1L Monoclonal antibody specifically detects PDIK1L in Human samples. It is validated for Western BlotSpecifications
| PDIK1L | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8N165 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 149420 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4G1 | |
| Unconjugated | |
| PDIK1L | |
| BC027088; casein kinase; CLIK1L; PDIK1L; PDLIM1 interacting kinase 1 like; PDLIM1-interacting kinase 1-like; Serine/threonine-protein kinase PDIK1L; STK35L2 | |
| Human recombinant protein fragment corresponding to amino acids 131-341 of human PDIK1L produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |