Learn More
PDIK1L Monoclonal Antibody (OTI4G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811104
This item is not returnable.
View return policy
Description
PDIK1L is a protein coding gene.Specifications
| PDIK1L | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8N165 | |
| PDIK1L | |
| Human recombit protein fragment corresponding to amino acids 131-341 of human PDIK1L produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4G1 | |
| Unconjugated | |
| PDIK1L | |
| CLIK1L, STK35L2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 149420 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |