missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDK-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 369.00 - € 529.00
Specifications
| Antigen | PDK-1 |
|---|---|
| Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625698
|
Novus Biologicals
NBP2-48631-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606788
|
Novus Biologicals
NBP2-48631 |
0.1 mL |
€ 529.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDK-1 Polyclonal antibody specifically detects PDK-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| PDK-1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, mTOR Pathway, Signal Transduction | |
| PBS (pH 7.2), 40% Glycerol | |
| 5170 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 3-phosphoinositide-dependent protein kinase 1, 3-phosphoinositide-dependent protein kinase 2 pseudogene, PDK1, PDPK2, PDPK2P, PkB kinase like gene 1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: HSLSASDTGLPQRSGSNIEQYIHDLDSNSFELDLQFSEDEKRLLLEKQAGGNPWHQFVENNLILKMGPVDKRKG | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title