missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579798
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Heart Tissue, Rat Skeletal Muscle Tissue, HELA Cell, M231 Cell, COLO320 Cell, SW620 Cell. IHC: Human Prostatic Cancer Tissue.
PDK1 (3-phosphoinositide dependent kinase-1) is a serine/threonine protein kinase localized to the plasma membrane. It plays a vital role in insulin-induced glycogen synthesis, PKC activation, and activation of the AGC subfamily of protein kinases. Plays an important role in cellular responses to hypoxia and is important for cell proliferation under hypoxia. Protects cells against apoptosis in response to hypoxia and oxidative stress.
Specifications
| PDK1 | |
| Polyclonal | |
| Unconjugated | |
| PDK1 | |
| [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 1, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 1, mitochondrial; B830012B01; D530020C15Rik; isoenzyme 1; kinase isoenzyme 1; lipoamide; mitochondrial pyruvate dehydrogenase, lipoamide, kinase isoenzyme 1; PDH kinase 1; PDHK1; PDK p48; Pdk1; pyruvate dehydrogenase; pyruvate dehydrogenase kinase 1; Pyruvate dehydrogenase kinase isoform 1; pyruvate dehydrogenase kinase, isoenzyme 1; pyruvate dehydrogenase kinase, isozyme 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116551, 5170 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| O15530, Q63065 | |
| PDK1 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human Mitochondrial Pyruvate dehydrogenase kinase 1 (408-421aa WKHYNTNHEADDWC). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction