missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDK4 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-07047UV
This item is not returnable.
View return policy
Description
PDK4 Polyclonal specifically detects PDK4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| PDK4 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 4, mitochondrial, EC 2.7.11, EC 2.7.11.2, FLJ40832, Pyruvate dehydrogenase kinase isoform 4, pyruvate dehydrogenase kinase, isoenzyme 4, pyruvate dehydrogenase kinase, isozyme 4, pyruvate dehydrogenase, lipoamide, kinase isozyme 4, mitochondrial | |
| Rabbit | |
| Affinity Purified | |
| Cancer, Lipid and Metabolism, Mitochondrial Markers, Protein Kinase, Signal Transduction | |
| 5166.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| DyLight 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PDK4 | |
| Synthetic peptide made to an internal portion of the human PDK4 protein (within residues 100-150). [Swiss-ProtNo. Q16654] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction