missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDLIM1 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98546DL594
This item is not returnable.
View return policy
Description
PDLIM1 Polyclonal specifically detects PDLIM1 in Human samples. It is validated for Western Blot.
Specifications
| PDLIM1 | |
| Polyclonal | |
| Western Blot | |
| CLIM1carboxyl terminal LIM domain protein 1, CLP-36, CLP36carboxy terminal LIM domain protein 1, C-terminal LIM domain protein 1, elfin, enigma, hCLIM1, LIM domain protein CLP-36, PDZ and LIM domain 1, PDZ and LIM domain 1 (elfin), PDZ and LIM domain protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human PDLIM1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 9124 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction