missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDLIM2 Monoclonal Antibody (OTI10C4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525505
This item is not returnable.
View return policy
Description
PDLIM2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Probable adapter protein located at the actin cytoskeleton that promotes cell attachment. Necessary for the migratory capacity of epithelial cells. Overexpression enhances cell adhesion to collagen and fibronectin and suppresses anchorage independent growth. May contribute to tumor cell migratory capacity.
Specifications
| PDLIM2 | |
| Monoclonal | |
| 0.44 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96JY6, Q9GKU1 | |
| PDLIM2 | |
| Full length human recombinant protein of PDLIM2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Canine, Cynomolgus Monkey | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI10C4 | |
| Unconjugated | |
| PDLIM2 | |
| 4732462F18Rik; hypothetical protein; mystique; Pdlim2; PDZ and LIM domain 2; PDZ and LIM domain 2 (mystique); PDZ and LIM domain protein 2; PDZ-LIM protein mystique; PP6345; Slim; STAT-interacting LIM protein | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 102126589, 486123, 64236 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction