missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDLIM2 Monoclonal Antibody (OTI15C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502874
This item is not returnable.
View return policy
Description
Probable adapter protein located at the actin cytoskeleton that promotes cell attachment. Necessary for the migratory capacity of epithelial cells. Overexpression enhances cell adhesion to collagen and fibronectin and suppresses anchorage independent growth. May contribute to tumor cell migratory capacity.Specifications
| PDLIM2 | |
| Monoclonal | |
| Unconjugated | |
| PDLIM2 | |
| MYSTIQUE, SLIM | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 64236 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot | |
| OTI15C1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96JY6 | |
| PDLIM2 | |
| Full length human recombit protein of human PDLIM2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction