Learn More
PDLIM2 Monoclonal Antibody (OTI15C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA502874S
Encompass_Preferred
 View more versions of this product
Product Code. 17587971
  • € 242.00 / 30µL
Description
PDLIM2 Monoclonal antibody specifically detects PDLIM2 in Human samples. It is validated for Flow Cytometry, Western Blot
Specifications
Specifications
PDLIM2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q15555
Mouse
Affinity chromatography
RUO
10982
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Flow Cytometry, Western Blot
OTI15C1
Unconjugated
MAPRE2
4732462F18Rik; hypothetical protein; mystique; Pdlim2; PDZ and LIM domain 2; PDZ and LIM domain 2 (mystique); PDZ and LIM domain protein 2; PDZ-LIM protein mystique; PP6345; Slim; STAT-interacting LIM protein
Full length human recombinant protein of human MAPRE2 produced in HEK293T cell.