missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA6 Monoclonal Antibody (OTI3B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800107S
This item is not returnable.
View return policy
Description
PSMA6 Monoclonal antibody specifically detects PSMA6 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| PSMA6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P60900 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5687 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3B8 | |
| Unconjugated | |
| PSMA6 | |
| 27 kDa prosomal protein; EGK_18113; IOTA; Macropain iota chain; macropain subunit iota; MGC22756; MGC2333; MGC23846; multicatalytic endopeptidase complex iota chain; p27K; PROS 27; PROS27; PROS-27; prosomal P27K protein; proteasome (prosome, macropain) subunit, alpha type 6; proteasome (prosome, macropain) subunit, alpha type, 6; proteasome 20S subunit alpha 6; proteasome alpha 6 subunit; proteasome iota chain; proteasome subunit alpha 6; proteasome subunit alpha type-6; proteasome subunit iota; PSA6; PSMA 6; PSMA6; testicular secretory protein Li 44 | |
| Full length human recombinant protein of human PSMA6 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction