Learn More
Invitrogen™ PDLIM3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA592915
Description
Immunogen sequence: KAAAHQLCLK IDRGETHLWS PQVSEDGKAH PFKINLESEP QEFKPIGTAH NRRAQPFVAA ANIDDKRQVV SASYNSPIGL YSTSNIQDAL HGQLRGLIPS SPQNEPTASV PPESDVYRML HDNRNEPTQP RQSGSFRVLQ GMVDDGSDDR PAGTRSVRAP V; Positive Samples: Mouse lung; Cellular Location: Cytoplasm, Z line, myofibril, sarcomere.
The protein encoded by this gene contains a PDZ domain and a LIM domain, indicating that it may be involved in cytoskeletal assembly. In support of this, the encoded protein has been shown to bind the spectrin-like repeats of alpha-actinin-2 and to colocalize with alpha-actinin-2 at the Z lines of skeletal muscle. This gene is found near a region of chromosome 4 that has been implicated in facioscapulohumeral muscular dystrophy, but this gene does not appear to be involved in the disease. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| PDLIM3 | |
| Polyclonal | |
| Unconjugated | |
| PDLIM3 | |
| actinin alpha 2 associated LIM protein; actinin-associated LIM protein; Actn2lp; AI463105; ALP; alpha-actinin-2-associated LIM protein; enigma homolog; PDLIM3; PDZ and LIM domain 3; PDZ and LIM domain protein 3; SK-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 53318 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O70209 | |
| PDLIM3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 70-230 of human PDLIM3 (NP_0011075791). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.