missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDLIM4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98063-100ul
This item is not returnable.
View return policy
Description
PDLIM4 Polyclonal antibody specifically detects PDLIM4 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| PDLIM4 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
| LIM domain protein, LIM protein RIL, PDZ and LIM domain 4, PDZ and LIM domain protein 4, Reversion-induced LIM protein, RILenigma homolog | |
| Produced in rabbits immunized with E. coli-derived Human PDLIM4 fragment. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 8572 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction