Learn More
PDLIM7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10221-1-AP-20UL
 View more versions of this product
Product Code. 16876463
  • € 145.00 / 20µL
Description
The protein encoded by this gene is representative of a family of proteins composed of conserved PDZ and LIM domains. LIM domains are proposed to function in protein-protein recognition in a variety of contexts including gene transcription and development and in cytoskeletal interaction. The LIM domains of this protein bind to protein kinases, whereas the PDZ domain binds to actin filaments. The gene product is involved in the assembly of an actin filament-associated complex essential for transmission of ret/ptc2 mitogenic signaling. The biological function is likely to be that of an adapter, with the PDZ domain localizing the LIM-binding proteins to actin filaments of both skeletal muscle and nonmuscle tissues. Alternative splicing of this gene results in multiple transcript variants.
Specifications
Specifications
PDLIM7
Polyclonal
Unconjugated
PDLIM7
ENIGMA, LIM mineralization protein, LMP, PDLIM7, PDZ and LIM domain 7 (enigma), PDZ and LIM domain protein 7, Protein enigma
Rabbit
Antigen Affinity Chromatography
RUO
9260
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9NR12
PDLIM7
PDLIM7 Fusion Protein Ag0264