missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDP1/PPAPDC2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 168.00
Specifications
| Antigen | PDP1/PPAPDC2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
PDP1/PPAPDC2 Polyclonal specifically detects PDP1/PPAPDC2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| PDP1/PPAPDC2 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| NP_982278 | |
| 403313 | |
| Antibody was raised against a 13 amino acid synthetic peptide from near the carboxy terminus human PPAPDC2. The immunogen is located within amino acids 170 - 220 of PPAPDC2. Amino Acid Squence: LVRRRRPAHNNQMD | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| bA6J24.6, EC 3.1.3, EC 3.1.3.-, FLJ46512, FLJ90191, MGC15483, PDP1, phosphatidic acid phosphatase type 2 domain containing 2, Phosphatidic acid phosphatase type 2 domain-containing protein 2, polyisoprenoid diphosphate phosphatase type 1, PPAP2 domain-containing protein 2, presqualene diphosphate phosphatase, PSDP | |
| PLPP6 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title