missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase A1/CPA1 Antibody (CPA1/2712), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79880DL594
This item is not returnable.
View return policy
Description
Carboxypeptidase A1/CPA1 Monoclonal specifically detects Carboxypeptidase A1/CPA1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Carboxypeptidase A1/CPA1 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CPA1 | |
| Recombinant full-length human Carboxypeptidase A1/CPA1 protein (Uniprot: P15085) | |
| 0.1 mL | |
| Mast Cell Markers | |
| Human | |
| IgG2c κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CPA1/2712 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, EC 3.4.17, EC 3.4.17.1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction