Learn More
Invitrogen™ PDPK1 Recombinant Rabbit Monoclonal Antibody (24GB5425)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557548
 View more versions of this product
Product Code. 30275078
  • € 580.00 / 100µL
Description
PDPK1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
PDPK1 (3 Phosphoinositide Dependent Protein Kinase 1) phosphorylates AGC kinases. PDPK1 activates conventional PKC and PKC zeta through phosphorylation of critical threonine residues in the activation loop. PDPK1 also phosphorylates Protein Kinase B (PKB) at threonine 308 in the presence of phosphatidylinositol-3,4,5-trisphosphate. Active Akt inactivates Glycogen Synthase Kinase 3 (GSK3), eventually leading to the dephosphorylation and activation of glycogen synthase, and the stimulation of glycogen synthesis. Because of the role that PDPK1 plays in insulin-induced glycogen synthesis and PKC activation, it is a potentially important target for metabolic drug research.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PDPK1
Recombinant Monoclonal
Unconjugated
Pdpk1
3-phosphoinositide dependent protein kinase 1; 3-phosphoinositide dependent protein kinase-1; 3-phosphoinositide-dependent protein kinase 1; 3-phosphoinositide-dependent protein kinase 2 pseudogene; HGNC:8809; hPDK1; isoenzyme 1; kinase isoenzyme 1; lipoamide; mPDK1; OTTHUMP00000174525; OTTHUMP00000205076; PDK1; PDPK1; PDPK2; PDPK2P; Pkb kinase; PkB kinase like gene 1; PkB-like 1; PRO0461; Protein kinase B kinase
Rabbit
Affinity chromatography
RUO
5170, 81745
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB5425
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O15530, O55173
Pdpk1
A synthesized peptide derived from human PDPK1 (aa 366-381).
100 μL
Primary
Human, Rat