missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDPR Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548323
This item is not returnable.
View return policy
Description
PDPR Polyclonal Antibody for IHC (P), Western Blot
Decreases the sensitivity of PDP1 to magnesium ions, and this inhibition is reversed by the polyamine spermine.
Specifications
| PDPR | |
| Polyclonal | |
| Unconjugated | |
| PDPR | |
| 4930402E16Rik; AU018056; Kiaa1990; mKIAA1990; PDP3; PDPR; pyruvate dehydrogenase phosphatase regulatory subunit; pyruvate dehydrogenase phosphatase regulatory subunit, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 319518, 55066 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q7TSQ8, Q8NCN5 | |
| PDPR | |
| KLH conjugated synthetic peptide between 821-850 amino acids from the C-terminal region of human PDPR. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction