missing translation for 'onlineSavingsMsg'
Learn More

PDRG1 Monoclonal Antibody (OTI1F5), TrueMAB™, OriGene

Product Code. 17580604
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17580604 100 μL 100µL
17500614 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17580604 Supplier Origene Technologies Supplier No. TA808878

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PDRG may play a role in chaperone-mediated protein folding (Potential).

Specifications

Antigen PDRG1
Applications Western Blot
Classification Monoclonal
Clone OTI1F5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PDRG1
Gene Accession No. Q9NUG6
Gene Alias C20orf126, PDRG
Gene Symbols PDRG1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 33-117 of human PDRG1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81572
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.