missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDSS2 Antibody (OTI1D12), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73323AF647
This item is not returnable.
View return policy
Description
PDSS2 Monoclonal specifically detects PDSS2 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| PDSS2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PDSS2 | |
| Full length recombinant protein of human PDSS2 (NP_065114) produced in HEK293T cells. | |
| 0.1 mL | |
| 57107.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI1D12 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| All-trans-decaprenyl-diphosphate synthase subunit 2, bA59I9.3, C6orf210decaprenyl-diphosphate synthase subunit 2, Candidate tumor suppressor protein, chromosome 6 open reading frame 210, Decaprenyl pyrophosphate synthase subunit 2, decaprenyl pyrophosphate synthetase subunit 2, DLP1hDLP1, EC 2.5.1.91, prenyl (decaprenyl) diphosphate synthase, subunit 2, subunit 2 of decaprenyl diphosphate synthase | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Monkey, Primate | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction