missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDSS2 Monoclonal Antibody (OTI1C11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503976
This item is not returnable.
View return policy
Description
The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.Specifications
| PDSS2 | |
| Monoclonal | |
| 0.87 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q86YH6 | |
| PDSS2 | |
| Full length human recombit protein of human PDSS2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C11 | |
| Unconjugated | |
| PDSS2 | |
| C6orf210, COQ10D3, DLP1, bA59I9.3, hDLP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57107 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction