missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDSS2 Monoclonal Antibody (OTI1D12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525785
This item is not returnable.
View return policy
Description
PDSS2 Monoclonal Antibody for Western Blot, IHC (P), Flow
The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.
Specifications
| PDSS2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86YH6 | |
| PDSS2 | |
| Full length human recombinant protein of PDSS2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D12 | |
| Unconjugated | |
| PDSS2 | |
| 5430420P03Rik; All-trans-decaprenyl-diphosphate synthase subunit 2; bA59I9.3; C6orf210; Candidate tumor suppressor protein; COQ10D3; decaprenyl diphosphate synthase subunit 2; decaprenyl pyrophosphate synthase subunit 2; decaprenyl pyrophosphate synthetase subunit 2; decaprenyl-diphosphate synthase subunit 2; DLP1; EGK_15212; Gm60; hDLP1; hypothetical protein LOC436624; kd; mDLP1; Pdss2; Plmp; prenyl (decaprenyl) diphosphate synthase, subunit 2; prenyl (solanesyl) diphosphate synthase, subunit 2; prenyl diphosphate synthase, subunit 2; prenyltransferase-like mitochondrial protein; RP11-59I9.3; solanesyl-diphosphate synthase subunit 2; subunit 2 of decaprenyl diphosphate synthase; zgc:92156 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57107 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction