Learn More
PDSS2 Mouse anti-Human, Clone: OTI1E9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF503973
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.Specifications
| PDSS2 | |
| Monoclonal | |
| Unconjugated | |
| PDSS2 | |
| DmelCG4186 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57107 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1E9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q86YH6 | |
| PDSS2 | |
| Full length human recombit protein of human PDSS2 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.