missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDX-1/IPF1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22150AF350
This item is not returnable.
View return policy
Description
PDX-1/IPF1 Polyclonal specifically detects PDX-1/IPF1 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PDX-1/IPF1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Glucose-sensitive factor, IDX-1GSF, Insulin promoter factor 1, insulin promoter factor 1, homeodomain transcription factor, Insulin upstream factor 1, IPF1pancreas/duodenum homeobox protein 1, Islet/duodenum homeobox-1, MODY4IUF1, pancreatic and duodenal homeobox 1, pancreatic-duodenal homeobox factor 1, PDX-1IPF-1, somatostatin transcription factor 1, Somatostatin-transactivating factor 1, STF-1IUF-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PDX1 | |
| A synthetic peptide made to a C-terminal portion of the human PDX1 protein (between residues 100-200) [UniProt P52945] | |
| 0.1 mL | |
| Lipid and Metabolism, Stem Cell Markers | |
| Human, Mouse, Canine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction