missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDX-1/IPF1 Antibody (OTI3H3), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73332CL1
This item is not returnable.
View return policy
Description
PDX-1/IPF1 Monoclonal antibody specifically detects PDX-1/IPF1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| PDX-1/IPF1 | |
| Monoclonal | |
| CoraFluor 1 | |
| Glucose-sensitive factor, IDX-1GSF, Insulin promoter factor 1, insulin promoter factor 1, homeodomain transcription factor, Insulin upstream factor 1, IPF1pancreas/duodenum homeobox protein 1, Islet/duodenum homeobox-1, MODY4IUF1, pancreatic and duodenal homeobox 1, pancreatic-duodenal homeobox factor 1, PDX-1IPF-1, somatostatin transcription factor 1, Somatostatin-transactivating factor 1, STF-1IUF-1 | |
| Full length human recombinant protein of human PDX1 (NP_000200) produced in E.coli. | |
| 0.1 mL | |
| Lipid and Metabolism, Stem Cell Markers | |
| 3651 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Immunohistochemistry, Western Blot | |
| OTI3H3 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction