missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDX1 Monoclonal Antibody (OTI3H3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807372S
This item is not returnable.
View return policy
Description
PDX1 Monoclonal antibody specifically detects PDX1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| PDX1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NS61 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 30819 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| OTI3H3 | |
| Unconjugated | |
| KCNIP2 | |
| DLDBP; E3BP; Glucose-sensitive factor; GSF; Idx1; IDX-1; Insulin promoter factor 1; insulin promoter factor 1, homeodomain transcription factor; insulin promotor factor 1; insulin upstream factor 1; Ipf1; IPF-1; Islet/duodenum homeobox 1; islet/duodenum homeobox-1; IUF1; IUF-1; MODY4; PAGEN1; Pancreas/duodenum homeobox protein 1; pancreatic and duodenal homeobox 1; pancreatic and duodenal homeobox gene 1; pancreatic-duodenal homeobox factor 1; PDHX; PDX1; pdx-1; somatostatin transactivating factor 1; somatostatin transcription factor 1; somatostatin-transactivating factor 1; Stf1; STF-1 | |
| Full length human recombinant protein of human KCNIP2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction