missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDXDC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101798
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PDXDC1.
PDXDC1 (Pyridoxal Dependent Decarboxylase Domain Containing 1) is a Protein Coding gene. Diseases associated with PDXDC1 include Autosomal Recessive Nonsyndromic Deafness 32 and Fetal Akinesia Deformation Sequence 1. Gene Ontology (GO) annotations related to this gene include pyridoxal phosphate binding and carboxy-lyase activity. An important paralog of this gene is HDC. [GeneCards].
Specifications
| PDXDC1 | |
| Polyclonal | |
| Unconjugated | |
| Pdxdc1 | |
| 2210010A19Rik; AA415817; KIAA0251; Kiaa0251-hp; LP8165; Pdxdc1; pyridoxal dependent decarboxylase domain containing 1; pyridoxal-dependent decarboxylase domain containing 1; pyridoxal-dependent decarboxylase domain-containing protein 1; RGD1562597 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23042, 304721, 94184 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6P996, Q99K01 | |
| Pdxdc1 | |
| A synthesized peptide derived from human PDXDC1(Accession Q6P996), corresponding to amino acid residues I676-I726. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction