missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDXK Monoclonal Antibody (OTI3G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503031S
This item is not returnable.
View return policy
Description
PDXK Monoclonal antibody specifically detects PDXK in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| PDXK | |
| Monoclonal | |
| 0.45 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P06730 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1977 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3G2 | |
| Unconjugated | |
| Eif4e | |
| 2310036D04Rik; AA119688; C21orf124; C21orf97; C77999; epididymis secretory sperm binding protein Li 1a; HEL-S-1a; Pdxk; Pkh; PNK; PRED79; pyridoxal (pyridoxine, vitamin B6) kinase; pyridoxal kinase; pyridoxamine kinase; pyridoxine kinase; vitamin B6 kinase | |
| Full length human recombinant protein of human EIF4E produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction