missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDXK Recombinant Rabbit Monoclonal Antibody (5N4M3)

Product Code. 30131750
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30131750 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30131750 Supplier Invitrogen™ Supplier No. MA555516

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen Sequence: MEEECRVLSI QSHVIRGYVG NRAATFPLQV LGFEIDAVNS VQFSNHTGYA HWKGQVLNSD ELQELYEGLR LNNMNKYDYV LTGYTRDKSF LAMVVDIVQE LKQQNPRLVY VCDPVLGDKW DGEGSMYVPE DLLPVYKEKV VPLADIITPN QFEAELLSGR KIHSQEEALR VMDMLHSMGP DTVVITSSDL PSPQGSNYLI VLGSQRRRNP AGSVVMERIR MDIRKVDAVF VGTGDLFAAM LLAWTHKHPN NLKVACEKTV STLHHVLQRT IQCAKAQAGE GVRPSPMQLE LRMVQSKRDI EDPEIVVQAT VL.

Pyridoxal kinase (PDXK) converts vitamin B6 to pyridoxal-5-phosphate (PLP), an essential cofactor in the intermediate metabolism of amino acids and neurotransmitters. The PDXK gene encodes a 312-amino acid polypeptide, and expression of the cDNA reveals pyridoxal kinase activity. Northern blot analysis revealed that a major 1.5-kb PDXK transcript is expressed in all tissues tested. The expression of PDXK shows circadian oscillations. The expression of Pdxk in mouse liver and brain is regulated by the 3 PAR bZIP transcription factors, Dbp, Hlf, and Tef, which also show circadian oscillations in expression. Mice devoid of all 3 transcription factors show decreased levels of brain PLP, serotonin, and dopamine, and are highly susceptible to frequently lethal generalized spontaneous and audiogenic epilepsies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDXK
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 5N4M3
Concentration 1.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene PDXK
Gene Accession No. O00764
Gene Alias 2310036D04Rik; AA119688; C21orf124; C21orf97; C77999; epididymis secretory sperm binding protein Li 1a; HEL-S-1a; Pdxk; Pkh; PNK; PRED79; pyridoxal (pyridoxine, vitamin B6) kinase; pyridoxal kinase; pyridoxamine kinase; pyridoxine kinase; vitamin B6 kinase
Gene Symbols PDXK
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-312 of human PDXK.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8566
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.