missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDXP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48487-25ul
This item is not returnable.
View return policy
Description
PDXP Polyclonal antibody specifically detects PDXP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PDXP | |
| Polyclonal | |
| Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
| chronophin, CIN, dJ37E16.5, EC 3.1.3, EC 3.1.3.74, FLJ32703, PLP, PLP phosphatase, PLPP, pyridoxal (pyridoxine, vitamin B6) phosphatase, pyridoxal phosphate phosphatase | |
| This antibody was developed against a recombinant protein corresponding to amino acids: FAKLREACAHLRDPECLLVATDRDPWHPLSDGSRTPGTGSLAAAVETASGRQALVVGKPSPYMFECITENFSIDPARTLMVGDRLETDILFGHRCGMTTVLTLTGVSRLEEAQAYLAAGQHDLVPHYYVESIADLTE | |
| 25 μL | |
| Cancer, Cell Biology, Cytoskeleton Markers, Endocrinology, Signal Transduction | |
| 57026 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction