missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDXP Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142628
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
Pyridoxal 5-prime-phosphate (PLP) is the active form of vitamin B6 that acts as a coenzyme in maintaining biochemical homeostasis. The preferred degradation route from PLP to 4-pyridoxic acid involves the dephosphorylation of PLP by PDXP. Pyridoxal 5-prime-phosphate (PLP) is the active form of vitamin B6 that acts as a coenzyme in maintaining biochemical homeostasis. The preferred degradation route from PLP to 4-pyridoxic acid involves the dephosphorylation of PLP by PDXP (Jang et al., 2003 ).
Specifications
| PDXP | |
| Polyclonal | |
| Unconjugated | |
| PDXP | |
| 1600027H05Rik; AB041662; chronophilin; chronophin; CIN; dJ37E16.5; FLJ32703; PDXP; Plp; PLP phosphatase; PLPP; PLP-phosophatase; pyridoxal (pyridoxine, vitamin B6) phosphatase; pyridoxal phosphatase; pyridoxal phosphate phosphatase; Rbp1; reg I binding protein I; reg I-binding protein 1; testicular secretory protein Li 36 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 57026, 57028, 727679 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P60487, Q8VD52, Q96GD0 | |
| PDXP | |
| Peptide with sequence C-QHDLVPHYYVES. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction