missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDZD11 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA580888

Product Code. 16337964

  • € 348.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human PDZD11/PDZK11.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PDZD11
Polyclonal
Unconjugated
PDZD11
AIPP1; ATPase-interacting PDZ protein; HSPC227; PDZ domain containing 11; PDZ domain-containing protein 11; PDZD11; PDZK11; PISP; Plasma membrane calcium ATPase-interacting single-PDZ protein; PMCA-interacting single-PDZ protein; UNQ6486/PRO21335
Rabbit
Antigen affinity chromatography, Protein A
RUO
51248
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunohistochemistry (Paraffin)
1 mg/mL
PBS with no preservative
Q5EBL8
PDZD11
Recombinant Human PDZD11/PDZK11 protein (Asp2-His140).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.