missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDZD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549762
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total PDZD2 protein.
Proteins containing PDZ domains have been shown frequently to bind the C-termini of transmembrane receptors or ion channels. They have also been shown to bind to other PDZ domain proteins and could possibly be involved in intracellular signaling. The protein encoded by this gene contains six PDZ domains and shares sequence similarity with pro-interleukin-16 (pro-IL-16). Like pro-IL-16, the encoded protein localizes to the endoplasmic reticulum and is thought to be cleaved by a caspase to produce a secreted peptide containing two PDZ domains. In addition, this gene is upregulated in primary prostate tumors and may be involved in the early stages of prostate tumorigenesis.
Specifications
| PDZD2 | |
| Polyclonal | |
| Unconjugated | |
| PDZD2 | |
| 4930537L06Rik; A930022H17Rik; activated in prostate cancer protein; AIPC; Gm82; KIAA0300; mKIAA0300; PAPIN; PDZ domain containing 2; PDZ domain containing 3; PDZ domain-containing protein 2; PDZ domain-containing protein 3; PDZD2; PDZK3; Pin1; Plakophilin-related armadillo repeat protein-interacting PDZ protein; plakophilin-related armadillo repeat protein-interactive PDZ protein; Processed PDZ domain-containing protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23037 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15018 | |
| PDZD2 | |
| Synthesized peptide derived from internal of human PDZD2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction