Learn More
VITLAB™ PE-HD Storage Bottles, With Tap
Click to view available options
Capacity (Metric):
5 L
10 L
25 L
Unit Size:
Each
Description
Durable transparent bottles made with high-density polyethylene (HDPE). With or without a polypropylene tap in a variety of capacities and mouth sizes.
- HDPE is characterized by a large strength-to-density ratio
- Transparent bottles allow for easy identification of contents
- Features a secure screw cap to protect the contents, even when the bottle is knocked over
- Stable carrying handles (two handles on the 25 and 50L sizes) ensure easy handling
- Available with easily-operated taps with a 3⁄4 in. pipe fitting for easy dispensing
- Wide-mouth bottles available for easy filling, especially of powder and paste-like substances
Specifications
Specifications
| Capacity (Metric) | 25 L |
| Color | Transparent |
| For Use With (Application) | Storing Liquids. |
| Includes | Tap, two carrying handles, screw cap |
| Material | HDPE |
| No. per Pack | 1Pack |
| Product Line | Storage Bottles, PE-HD, With Tap |
| Thread Size (Metric) | 95.5mm |
| Mouth | Narrow |
| Height (Metric) | 565 mm |
| Show More |