Learn More
VITLAB™ PE-HD Storage Bottles, Without Tap
Click to view available options
Capacity (Metric):
5 L
10 L
Mouth:
Narrow
Wide
Thread Size (Metric):
62.5mm
94mm
108mm
Unit Size:
Each
Description
Durable transparent bottles made with high-density polyethylene (HDPE). With or without a polypropylene tap in a variety of capacities and mouth sizes.
- HDPE is characterized by a large strength-to-density ratio
- Transparent bottles allow for easy identification of contents
- Features a secure screw cap to protect the contents, even when the bottle is knocked over
- Stable carrying handles (two handles on the 25 and 50L sizes) ensure easy handling
- Available with easily-operated taps with a 3⁄4 in. pipe fitting for easy dispensing
- Wide-mouth bottles available for easy filling, especially of powder and paste-like substances
Specifications
Specifications
| Capacity (Metric) | 5 L |
| Color | Transparent |
| For Use With (Application) | Storing Liquids. |
| Includes | Carrying handle, screw cap |
| Material | HDPE |
| No. per Pack | 1Pack |
| Product Line | Storage Bottles, PE-HD, Without Tap |
| Thread Size (Metric) | 62.5mm |
| Mouth | Narrow |
| Height (Metric) | 345 mm |
| Show More |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido